Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18610187 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-187 200 L Drum
18-610-186 100 mL Amber Glass Bottle
18-610-188 500 mL Amber Glass Bottle
18-610-184 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-187 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1250200LTBL

Chemical Identifiers

CAS 75-09-2
Molecular Weight (g/mol) 84.93

Specifications

CAS Max % 1
Quantity 200 L
Identification To Pass Test
Packaging Drum
Residue after Evaporation 2e-05
Titratable Acid 0.0003 meq/g
Water 0.0002
Boiling Point 39.8°C
Percent Purity 0.995
Grade Reagent
Chemical Name or Material Dichloromethane
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.