Learn More
Dichloromethane, Purified (Methylene Chloride), Reagents
Specifications
Specifications
| Boiling Range | 39.8°C |
| Melting Point | -96.7°C |
| Color | Colorless |
| Boiling Point | 39.8°C |
| Melting Range | -96.7°C |
| Packaging | Steel Pail |
| Quantity | 20 L |
| Specific Gravity | 1.3266 |
| Grade | Reagent |
| Physical Form | Liquid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.