Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99.9% (GC), Solstice

Catalog No. 60046093 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
4 x 2.5 L
6 x 1 L
250 kg
Packaging:
Glass Bottle
Metal Drum
Stainless Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-093 1 L Glass Bottle
60-046-099 6 x 1 L Glass Bottle
60-046-098 4 x 2.5 L Glass Bottle
60-046-091 10 L Stainless Steel Can
60-046-095 250 kg Metal Drum
60-046-094 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-046-093 Supplier Solstice Supplier No. 322221L

Puriss. p.a., ACS Reagent, Reag. ISO,≥99.9% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Appearance Liquid
Density 1.325g/mL at 25°C, 1.320 to 1.330g/Lat 20°C
Quantity 1 L
Explosion Limit 0.22
Ignition Point 1223°F
Packaging Glass Bottle
Refractive Index n20/D 1.4235 to 1.4245
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid: ≤0.001% (as HCl)
Vapor Pressure 6.83psi (20°C), 24.45psi (55°C)
UN Number UN1593
Beilstein 1730800
Boiling Point 39.8°C to 40°C
Formula Weight 84.93g/mol
Percent Purity ≥99.9% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Dichloromethane, Reag. ISO
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
Precautionary Statement H315-H319-H335-H336-H351-H373
DOTInformation 6.1/PGIII
EINECSNumber 200-838-9
RTECSNumber PA8050000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.