Learn More
Medchemexpress LLC HY-12638 500mg Medchemexpress, Dichlorophen CAS:97-23-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12638500MG
Encompass_Preferred
Medchemexpress, HY-12638 500mg Dichlorophen CAS:97-23-4 Dichlorophen (DDM) is an anticestodal agent. Dichlorophen is an antimicrobial agent shown to exert activity against cestodes, protozoa, fungi, and bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1099
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.