Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorophenylborane 98.0+%, TCI America™
SDP

Catalog No. D436725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D436725G 25 g
D43675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D436725G Supplier TCI America Supplier No. D436725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 873-51-8
Molecular Formula C6H5BCl2
Molecular Weight (g/mol) 158.816
MDL Number MFCD00013612
InChI Key NCQDQONETMHUMY-UHFFFAOYSA-N
Synonym phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron
PubChem CID 136678
IUPAC Name dichloro(phenyl)borane
SMILES B(C1=CC=CC=C1)(Cl)Cl

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 177°C
Quantity 25 g
UN Number 2924
Formula Weight 158.82
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Dichlorophenylborane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.