Learn More
AdipoGen Diclofop-methyl
SDP
Supplier:  AdipoGen CDXD0867M500
Encompass_Preferred
Chemical. CAS 51338-27-3. Formula C16H14Cl2O4. MW 341.19. Diclofop-methyl is an early post-emergence phenoxypropanoic acid herbicide. Its mode of action involves inhibition of acetyl CoA carboxylase ACCase activity, which in turn decreases fatty acid synthesis in weeds and inhibits photosynthesis and the activity of meristem. Can be used as a reference compound.
Catalog No. 50-256-1031
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.