Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dicyclohexano-18-Crown-6

Catalog No. 15840210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-840-210G 10 g
15-840-21G 1 g
15-840-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 15-840-210G Supplier Sigma Aldrich Supplier No. 15840210G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Anion activator, and complexing agent which solubilizes alkali metal ions in non-polar solvents.

Specifications

CAS 16069-36-6
Molecular Formula C20H36O6
Linear Formula C20H36O6
MDL Number MFCD00005099
Quantity 10 g
Synonym 2,3,11,12-Dicyclohexano-1,4,7,10,13,16-hexaoxacyclooctadecane; 2,5,8,15,18,21-Hexaoxatricyclo[20.4.0.09.14]hexacosane; Dicyclohexyl-18-crown-6; Perhydro-dibenzo-18-crown-6
Molecular Weight (g/mol) 372.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.