Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexano-18-crown 6-Ether 98.0+%, TCI America™
SDP

Catalog No. D16681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16681G 1 g
D16685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16681G Supplier TCI America Supplier No. D16681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16069-36-6
Molecular Formula C20H36O6
Molecular Weight (g/mol) 372.502
MDL Number MFCD00005099
InChI Key BBGKDYHZQOSNMU-UHFFFAOYSA-N
Synonym dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
PubChem CID 85955
SMILES C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

Specifications

Melting Point 51°C
Color White
Boiling Point 344°C
Quantity 1 g
UN Number 2811
Formula Weight 372.50
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Dicyclohexano-18-crown 6-Ether
RTECSNumber HP5385000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.