Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine 97.0+%, TCI America™
SDP

Catalog No. D51351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51351G 1 g
D5135200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51351G Supplier TCI America Supplier No. D51351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 303111-96-8
Molecular Formula C27H45P
Molecular Weight (g/mol) 400.63
MDL Number MFCD11656073
InChI Key DTSPXGRQYHLKLO-UHFFFAOYSA-N
Synonym dicyclohexyl 2,4,6-triisopropylphenyl phosphine, 2,4,6 tri-isopropyl phenyl di-cyclohexylphosphine, 2,4,6-tri-isopropyl phenyl di-cyclohexylphosphine, dicyclohexyl 2,4,6-tris 1-methylethyl phenyl phosphine, dicyclohexyl 2,4,6-triisopropylphenyl phosphane, acmc-1ajbl, 2,4,6 triisopropyl phenyl dicyclohexylphosphine, 1,3,5-triisopropyl-2-dicyclohexylphosphino benzene, dicyclohexyl 2,4,6-tri propan-2-yl phenyl phosphane
PubChem CID 9801140
IUPAC Name dicyclohexyl[2,4,6-tris(propan-2-yl)phenyl]phosphane
SMILES CC(C)C1=CC(C(C)C)=C(P(C2CCCCC2)C2CCCCC2)C(=C1)C(C)C

Specifications

Melting Point 139°C
Color White
Quantity 1 g
Formula Weight 400.63
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.