Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didecyl Phosphate 95.0+%, TCI America™
SDP

Catalog No. P10111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10111G 1 g
P1011100MG 100 mg
P10115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P10111G Supplier TCI America Supplier No. P10111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7795-87-1
Molecular Formula C20H43O4P
Molecular Weight (g/mol) 378.53
MDL Number MFCD00015062
InChI Key QHAUASBJFFBWMY-UHFFFAOYSA-N
Synonym Phosphoric Acid Didecyl Ester
PubChem CID 82260
IUPAC Name bis(decyloxy)phosphinic acid
SMILES CCCCCCCCCCOP(O)(=O)OCCCCCCCCCC

Specifications

Melting Point 47°C
Color White
Quantity 1 g
Formula Weight 378.53
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Didecyl Phosphate
EINECSNumber (2)-1986
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.