Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didecyl phthalate, 95%

Catalog No. AC407101000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407101000 100 g Glass bottle
AC407100250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407101000 Supplier Thermo Scientific Chemicals Supplier No. 407101000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 84-77-5 | C28H46O4 | 446.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-77-5
Molecular Formula C28H46O4
Molecular Weight (g/mol) 446.67
MDL Number MFCD00041915
InChI Key PGIBJVOPLXHHGS-UHFFFAOYSA-N
Synonym didecyl phthalate, decyl phthalate, di-n-decyl phthalate, vinicizer 105, phthalic acid, didecyl ester, vinysize 105, 1,2-benzenedicarboxylic acid, didecyl ester, bis n-decyl phthalate, didecyl 1,2-benzenedicarboxylate, unii-fi5fbn947z
PubChem CID 6788
ChEBI CHEBI:34676
IUPAC Name didecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCC

Specifications

Melting Point 4.0°C
Density 0.9700g/mL
Boiling Point 261.0°C (5.0 mmHg)
Flash Point 232°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4790 to 1.4840
Quantity 100 g
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Formula Weight 446.66
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Didecyl phthalate, 95%
Show More Show Less
EINECSNumber 201-561-6
RTECSNumber TI0900000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.