Learn More
TARGETMOL CHEMICALS INC DIDESMETHYL CARIPRAZINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T15117
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Didesmethyl cariprazine a metabolite of Cariprazine is the major cyclic active part. Didesmethyl cariprazine has a long half-life of 1-3 weeks. Cariprazine is a potent D3 preference D3/D2 receptor partial agonist of dopamine for the treatment of schizophrenia bipolar mania and depression with a high affinity for d3 and d2 receptors and a moderate affinity for 5-HT 1A receptors. purity: 99%
Catalog No. 50-258-1252
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.