Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dieldrin Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USPP1901
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-1901 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PP-1901 Supplier Agilent Technologies Supplier No. PP1901
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-57-1 , 67-56-1
Molecular Formula C12H8Cl6O
Molecular Weight (g/mol) 380.895
InChI Key DFBKLUNHFCTMDC-YNXGVQRSSA-N
PubChem CID 101820673
SMILES C1C2C3C(C1C4C2O4)C5(C(=C(C3(C5(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,dieldrin
Element Methanol,dieldrin
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Dieldrin Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.