Learn More
Abcam Dienogest, Steroidal Progesterone, Solid, >99% Purity, 100 mg
Description
- Molecular Weight: 311.4 Da
- Purity: >99%
- Description: Steroidal progesterone; non-ethinylated progestin structurally related to testosterone
- Activity: Anti-androgenic and antiproliferative effects in vitro
- Form: Solid
- CAS Number: 65928-58-7
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Form | Solid |
| Target Molecule | Dienogest |
| Buffer | 100% Dienogest |
| Quantity | 100 mg |
| For Use With (Application) | In vitro studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.