Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dienogest, Steroidal Progesterone, Solid, >99% Purity, 25 mg

Catalog No. 502706713
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6713 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6713 Supplier Abcam Supplier No. AB14128225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-ethinylated progestin with anti-androgenic and antiproliferative effects in vitro.

  • Molecular weight: 311.4 Da
  • Purity: >99%
  • Description: Steroidal progesterone, non-ethinylated progestin structurally related to testosterone
  • Biological activity: Anti-androgenic and antiproliferative effects in vitro
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Dienogest
Shipping Condition Ambient
Buffer 100% Dienogest
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Dienogest
Quantity 25 mg
For Use With (Application) Anti-androgenic and antiproliferative research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.