Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dienogest 98.0+%, TCI America™
SDP

Catalog No. D5230250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5230250MG 250 mg
D523050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5230250MG Supplier TCI America Supplier No. D5230250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65928-58-7
Molecular Formula C20H25NO2
Molecular Weight (g/mol) 311.425
MDL Number MFCD00868356
InChI Key AZFLJNIPTRTECV-FUMNGEBKSA-N
Synonym 17alpha-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Cyanomethyl-17-hydroxy-estra-4,9-dien-3-one
PubChem CID 68861
ChEBI CHEBI:70708
IUPAC Name 2-[(8S,13S,14S,17R)-17-hydroxy-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]acetonitrile
SMILES CC12CCC3=C4CCC(=O)C=C4CCC3C1CCC2(CC#N)O

Specifications

Melting Point 215°C
Color White
Quantity 250 mg
Formula Weight 311.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dienogest
RTECSNumber RC8899500
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.