Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diesel in Water, PT Concentrate for dilution to 1 L, Proficiency Testing Material, MilliporeSigma™ Supelco™

Catalog No. 111017368 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7368 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7368 Supplier MilliporeSigma Supelco Supplier No. PE17082ML

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

This ampule contains a concentrate of a No. 2 Diesel in methanol. This PT sample is packaged as a 1.5 mL concentrate. The solvent for this sample is methanol. The PT sample is not preserved. Sample preparationTo prepare solution, fill a 1000-mL volumetric flask to volume with carbon-filtered reagent-grade water at room temperature. Open ampule PE1708-2ML (be careful to avoid cuts on glass) and immediately withdraw 1 mL. Quickly inject the 1 mL aliquot into the reagent water in the expanded area of the volumetric flask. Mix the sample by inverting the flask three times only and analyze immediately. Note: This sample may be used for method AK102 SV (small volume). To ensure sample integrity, we recommend preparing the same small volume as you normally test. For instance, if your small volume sample is 125 mL, the PT should be diluted 125 uL of concentrate to 125 mL of water (1:1000 dilution).
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) Diesel Range Organics (DRO) 800-6000 μg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.