Learn More
Streck Diesse MINI-CUBE Transponder
SDP
Catalog No. 23285253 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
1000 Tests
500 Tests
5000 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-285-253 5000 Tests
23-285-258 500 Tests
23-285-259 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-285-253 Supplier Streck Supplier No. 240419
Add to Cart
Add to Cart
Request a Quote
A Diesse MINI-CUBE accessory used to load a pre-determined number of tests on the Diesse MINI-CUBE ESR Analyzer.
  • Diesse MINI-CUBE accessory used to load a pre-determined number of tests on the Diesse MINI-CUBE ESR Analyzer.
Specifications
Type ESR Analyzer
For Use With (Equipment) MINI-CUBE ESR Analyzer
Quantity 5000 Tests