Learn More
Sigma Organic Chemistry Diethanolamine | 100G | 111-42-2 | MFCD00002843
Supplier: Sigma Organic Chemistry D8885100G

Diethanolamine, 100G
About This Item:
Synonym(s): 2,2'-Iminodiethanol; Bis(2-hydroxyethyl)amine
Linear Formula: HN(CH2CH2OH)2
Molecular Weight: 105.14
CAS Number: 111-42-2
MDL Number: MFCD00002843
UNSPSC Code: 12352100
Purity: >=98.0%
Boiling Point: 217 C/150 mmHg (lit.)
Melting Point: 28 C (lit.)
Density: 1.097 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.477 (lit.)
Storage: room temp
EINECS Number: 203-868-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.