Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, 99%

Catalog No. AC113920250
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
Glass Bottle
Metal drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113920250 25 kg Metal drum
AC113920010 1 kg Glass Bottle
AC113920025 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC113920250 Supplier Thermo Scientific Chemicals Supplier No. 113920250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Melting Point 27.0°C to 30.0°C
Density 1.0955g/mL
Boiling Point 268.0°C
Flash Point 179°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Metal drum
Refractive Index 1.4750 to 1.4790
Linear Formula (HOCH2CH2)2NH
Quantity 25 kg
Beilstein 04, 283
Merck Index 15, 3118
Vapor Pressure 2hPa at 10°C
Formula Weight 105.14
Percent Purity 99%
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Diethanolamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. Harmful to aquatic life with long lasting effects. Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Take off contaminated clothing and wash before reuse. IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.