Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, Reagent, ACS, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18602183 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-183 100 mL Amber Glass Bottle
18-602-184 500 mL Amber Glass Bottle
18-602-185 6 x 500 mL Amber Glass Bottle
18-602-186 4 L Amber Glass Bottle
18-602-187 4 x 4 L Amber Glass Bottle
18-602-188 20 L Amber Glass Bottle
18-602-189 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-602-183 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1033100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

D1033, 111-42-2, C4H11NO2

Spectrum™ Chemical Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

CAS Max % 100%
Boiling Point >250°C
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 98.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material Diethanolamine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.