Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, 95%

Catalog No. AC399190010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399190010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399190010 Supplier Thermo Scientific Chemicals Supplier No. 399190010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1149-23-1 | C13H19NO4 | 253.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-23-1
Molecular Formula C13H19NO4
Molecular Weight (g/mol) 253.3
InChI Key LJXTYJXBORAIHX-UHFFFAOYSA-N
Synonym diludine, diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, hantzsch ester, ethidine, etidin, diethone, hantzsch's dihydropyridine, diethyl 2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, diludin, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-, diethyl ester
PubChem CID 70849
IUPAC Name diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1)C(=O)OCC)C)C

Specifications

Melting Point 178.0°C to 183.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 253.3
Percent Purity 95%
Chemical Name or Material Diethyl 1, 4-dihydro-3, 5-pyridinecarboxylate
Hazard Category GHS Signal Word: Warning
EINECSNumber 214-561-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.