Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl(3-pyridyl)borane, 97%, Thermo Scientific Chemicals

Catalog No. AC457750050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457750050 5 g
AC457750250 25 g
AC457751000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457750050 Supplier Thermo Scientific Chemicals Supplier No. 457750050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89878-14-8
Molecular Formula C9H14BN
Molecular Weight (g/mol) 147.03
MDL Number MFCD00012348
InChI Key OJKBCQOJVMAHDX-UHFFFAOYSA-N
Synonym diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo
PubChem CID 642851
IUPAC Name diethyl(pyridin-3-yl)borane
SMILES CCB(CC)C1=CN=CC=C1

Specifications

Melting Point 149.0°C to 151.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 147.03
Percent Purity 97%
Chemical Name or Material Diethyl(3-pyridyl)borane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.