Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (4-Chlorobenzyl)phosphonate 97.0+%, TCI America™
SDP

Catalog No. D333525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D333525G 25 g
D33355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D333525G Supplier TCI America Supplier No. D333525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39225-17-7
Molecular Formula C11H16ClO3P
Molecular Weight (g/mol) 262.67
InChI Key BVQPVBZRJSFOEZ-UHFFFAOYSA-N
Synonym diethyl 4-chlorobenzylphosphonate, diethyl 4-chlorobenzyl phosphonate, diethyl 4-chlorophenyl methylphosphonate, 1-chloro-4-diethoxyphosphorylmethyl benzene, phosphonic acid, 4-chlorophenyl methyl-, diethyl ester, phosphonic acid, p-4-chlorophenyl methyl-, diethyl ester, acmc-1aifv, diethyl 4-chlorobenzylposphonate, diethyl 4-chlorobenzylphosphate
PubChem CID 99240
IUPAC Name 1-chloro-4-(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)Cl)OCC

Specifications

Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 180°C
Formula Weight 262.67
Percent Purity ≥97.0% (GC)
Chemical Name or Material Diethyl (4-Chlorobenzyl)phosphonate
EINECSNumber (3)-3977
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.