Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (4-Cyanobenzyl)phosphonate 98.0+%, TCI America™
SDP

Catalog No. D332325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D332325G 25 g
D33235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D332325G Supplier TCI America Supplier No. D332325G

Chemical Identifiers

CAS 1552-41-6
Molecular Formula C12H16NO3P
Molecular Weight (g/mol) 253.24
MDL Number MFCD08274488
InChI Key MFXWOYIWKNJHPC-UHFFFAOYSA-N
Synonym (4-Cyanobenzyl)phosphonic Acid Diethyl Ester, 4-(Diethylphosphonomethyl)benzonitrile
PubChem CID 241438
IUPAC Name diethyl [(4-cyanophenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC=C(C=C1)C#N)OCC

Specifications

Melting Point 31°C
Color White-Yellow
Boiling Point 169°C
Quantity 25 g
UN Number 3439
Formula Weight 253.24
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Diethyl (4-Cyanobenzyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.