Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (4-Fluorobenzyl)phosphonate 98.0+%, TCI America™
SDP

Catalog No. D332425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D332425G 25 g
D33245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D332425G Supplier TCI America Supplier No. D332425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63909-58-0
Molecular Formula C11H16FO3P
Molecular Weight (g/mol) 246.22
MDL Number MFCD06795966
InChI Key FIYRZOAUPPNGAO-UHFFFAOYSA-N
Synonym (4-Fluorobenzyl)phosphonic Acid Diethyl Ester
PubChem CID 11064685
IUPAC Name diethyl [(4-fluorophenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC=C(F)C=C1)OCC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 246.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Fluorobenzyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.