Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Acetylenedicarboxylate 96.0+%, TCI America™
SDP

Catalog No. A0089250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0089250ML 250 mL
A008925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0089250ML Supplier TCI America Supplier No. A0089250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 762-21-0
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00009186
InChI Key STRNXFOUBFLVIN-UHFFFAOYSA-N
Synonym diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate
PubChem CID 69803
IUPAC Name diethyl but-2-ynedioate
SMILES CCOC(=O)C#CC(=O)OCC

Specifications

Melting Point 1°C
Color Yellow
Boiling Point 115°C
Quantity 250 mL
UN Number 1760
Formula Weight 170.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Acetylenedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.