Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (bromodifluoromethyl)phosphonate, 97%

Catalog No. AAL1671506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1671506 5 g
AAL1671503 1 g
AAL1671514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1671506 Supplier Thermo Scientific Chemicals Supplier No. L1671506
Add to Cart

CAS: 65094-22-6 | C5H10BrF2O3P | 267.01 g/mol

Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65094-22-6
Molecular Formula C5H10BrF2O3P
Molecular Weight (g/mol) 267.01
MDL Number MFCD00069118
InChI Key QRADKVYIJIAENZ-UHFFFAOYSA-N
Synonym diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7p
PubChem CID 144101
IUPAC Name 1-[[bromo(difluoro)methyl]-ethoxyphosphoryl]oxyethane
SMILES CCOP(=O)(OCC)C(F)(F)Br

Specifications

Density 1.503
Boiling Point 99°C to 102°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.416
Quantity 5 g
Beilstein 1944045
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 267.01
Percent Purity 97%
Chemical Name or Material Diethyl (bromodifluoromethyl)phosphonate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.