Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Chlorophosphate 97.0+%, TCI America™
SDP

Catalog No. D2206250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2206250G 250 g
D220625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2206250G Supplier TCI America Supplier No. D2206250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 814-49-3
Molecular Formula C4H10ClO3P
Molecular Weight (g/mol) 172.545
MDL Number MFCD00009075
InChI Key LGTLXDJOAJDFLR-UHFFFAOYSA-N
Synonym diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
PubChem CID 13139
IUPAC Name 1-[chloro(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)Cl

Specifications

Color Yellow
Boiling Point 93°C
Quantity 250 g
UN Number 2927
Formula Weight 172.54
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Diethyl Chlorophosphate
RTECSNumber TD1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.