Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Diselenide 96.0+%, TCI America™
SDP

Catalog No. D47151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47151G 1 g
D47155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47151G Supplier TCI America Supplier No. D47151G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 628-39-7
Molecular Formula C4H10Se2
Molecular Weight (g/mol) 216.066
MDL Number MFCD00015491
InChI Key OMAWWKIPXLIPDE-UHFFFAOYSA-N
Synonym Ethyl Diselenide
PubChem CID 69405
IUPAC Name (ethyldiselanyl)ethane
SMILES CC[Se][Se]CC

Specifications

Boiling Point 85°C
Quantity 1 g
UN Number 3440
Formula Weight 216.04
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Diselenide
RTECSNumber JM9151750
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.