Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph., ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Metal drum
60-29-7
C4H10O
74.123
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.123
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
For analysis
Acetone 0.002% max.
Acidity Passes Test
Aldehyde 0.0001% max.
Assay Percent Range 99.5%
Linear Formula (CH3CH2)2O
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
Cadmium (Cd) 0.02ppm max.
Formula Weight 74.12
Physical Form Liquid
Percent Purity 99.5%
Grade Analysis
Cobalt (Co) 0.02ppm max.
Copper (Cu) 0.02ppm max.
Density 0.7140g/mL
Ethanol 0.01% max.
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.05ppm max.
Methanol 0.01% max.
Nickel (Ni) 0.02ppm max.
Packaging Glass Bottle
Peroxides 1ppm max.
Refractive Index 1.3510 to 1.3530
Residue after Evaporation 0.002% max.
Solubility Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
Specific Gravity 0.714
Stabilizer 5ppm BHT
Water 0.1% max. (K.F.)
Flash Point −45°C
Viscosity 0.2448 mPa.s (20°C)
Boiling Point 34.6°C
Color Undesignated
Melting Point -116.0°C
Quantity 2.5L
Chemical Name or Material Diethyl ether, Stabilized with BHT
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger