Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018347 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8347 10 mL
11-102-0382 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8347 Supplier MilliporeSigma Supelco Supplier No. 9123810MLF
Add to Cart
Add to Cart

Diethyl ether is a colorless and highly volatile organic compound, which commonly finds applications as an extraction solvent.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-29-7
Ignition Point 320°F
Solution Contains ˜0.0005% 2,6-di- tert. -butyl-4-methylphenol as stabilizer
Vapor Pressure 2311 hPa (60°C)
Vapor Density 2.6 (vs. air)
Density 0.706 g/mL (at 25°C)
Flash Point -40°C
Physical Form Neat
Linear Formula (CH3 CH2)2 O
MDL Number MFCD00011646
Melting Point -116°C
Boiling Point 34.6°C
Synonym Ether; Ethyl ether
Type Analytical Standard
Quantity 10 mL
Formula Weight 74.12
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.