Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, ACS, 98% min, stab. with 0.001% BHT and 3% ethanol

Catalog No. AA3322436
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3322436 500 g
AA3322462 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3322436 Supplier Thermo Scientific Chemicals Supplier No. 03322436
Add to Cart

missing translation for 'validMainframeMsg'

Widely used as a solvent, dissolves P, S, Br2 and I2 | CAS: 60-29-7 | C4H10O | 74.12 g/mol

Widely used as a solvent. Dissolves P, S, Br2 and I2. It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point °C to 116°C
Density 0.713
Quantity 500 g
Flash Point -45°C (-49°F)
Physical Form Liquid
Refractive Index 1.3525
Assay Percent Range 98% min
Linear Formula (C2H5)2O
UN Number UN1155
Merck Index 14,3806
Boiling Point 34.6°C
Solubility Information Immiscible with water,miscible with alcohol.
Formula Weight 74.12
Percent Purity ≤98%
Grade ACS
Chemical Name or Material Diethyl ether, 0.7134, 1.35555
Show More Show Less
Hazard Category H224-H302-H305-H335-H336-H373
Hazard Statement GHS H Statement
H224-H302-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER
EINECSNumber 200-467-2
RTECSNumber KI5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.