Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%, Solstice
GSA_VA

Catalog No. 6000519 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-005-19 4 x 4 L Glass Bottle
60-005-18 1 L Glass Bottle
60-005-20 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-005-19 Supplier Solstice Supplier No. 3099664X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC, inhibitor-free,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
Absorbance max. 1.00 at 218 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 250 nm
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Flash Point −45°C
Packaging Glass Bottle
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34°C to 35°C
Formula Weight 74.12
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
EINECSNumber 200-467-2
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.