Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Ether For gas chromatography ECD and FID SupraSolv™ MilliporeSigma™

Catalog No. M1009311000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1009311000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1009311000 Supplier MilliporeSigma Supplier No. 1.00931.1000
Add to Cart
Add to Cart

For gas chromatography ECD and FID SupraSolv™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.3°C
Color Colorless
Density 0.71g/cm3 at 20°C
Quantity 1 L
Flash Point −40°C (−40°F)
Ignition Point 180°C (356°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 587 hPa (20°C)
UN Number 1155
Boiling Point 34.6°C
Solubility Information Soluble in water: 69g/L at 20°C
Formula Weight 74.12
Chemical Name or Material Diethyl Ether
Show More Show Less
Hazard Category H224: Extremely flammable liquid and vapour.
Hazard Statement H302: Harmful if swallowed.
Precautionary Statement H336: May cause drowsiness or dizziness.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.