Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, For pesticide residue analysis, Solstice

Catalog No. 6000524 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-005-24 6 x 1 L
60-005-25 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-005-24 Supplier Solstice Supplier No. 316716X1L
Add to Cart
Add to Cart

CHROMASOLV™ for pesticide residue analysis,≥99.8%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Color Colorless
Density 0.706g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.365
Flash Point −40°C (−40°F)
Ignition Point 320°F
Linear Formula (CH3CH2)2O
Vapor Pressure 2311 hPa (60°C)
UN Number 1155
Beilstein 1696894
Formula Weight 74.12g/mol
Grade CHROMASOLV
Chemical Name or Material Diethyl Ether
Show More Show Less
Hazard Statement P210-P261
Precautionary Statement H224-H302-H336
EINECSNumber 200-467-2
RTECSNumber KI5775000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.