Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, For pesticide residue analysis, Solstice
GSA_VA

Catalog No. 6000525 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-005-25 4 x 4 L
60-005-24 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-005-25 Supplier Solstice Supplier No. 316714X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ for pesticide residue analysis,≥99.8%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
Density 0.706 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Flash Point −45°C
Packaging Glass Bottle
Linear Formula (CH3CH2)2O
Water <0.05%
UN Number UN1155
Boiling Point 34°C to 35°C
Formula Weight 74.12
Percent Purity >99.8%
Grade Pesticide
Assay GC product+ethanol
Chemical Name or Material Diethyl Ether, CHROMASOLVä, for pesticide residue analysis
Show More Show Less
ShelfLife 1080 days from date of manufacture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.