Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl fumarate, 98%

Catalog No. AC114380010
Change view
Click to view available options
Quantity:
10 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114380010 1 kg Glass bottle
AC114380100 10 g Glass bottle
AC114382500 250 g Glass bottle
AC114380025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC114380010 Supplier Thermo Scientific Chemicals Supplier No. 114380010
Add to Cart

CAS: 623-91-6 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-91-6
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00064455
InChI Key IEPRKVQEAMIZSS-AATRIKPKSA-N
Synonym diethyl fumarate, fumaric acid, diethyl ester, anti-psoriaticum, diethyl 2e-but-2-enedioate, fumaric acid diethyl ester, diethyl e-but-2-enedioate, 2-butenedioic acid e-, diethyl ester, trans-2-butenedioic acid diethyl ester, 2-butenedioic acid 2e-, diethyl ester, unii-5wbu5a3e8a
PubChem CID 638144
ChEBI CHEBI:87388
IUPAC Name diethyl (E)-but-2-enedioate
SMILES CCOC(=O)\C=C\C(=O)OCC

Specifications

Melting Point 1.0°C to 2.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 218.0°C to 219.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4396 to 1.4416
Linear Formula C2H5O2CCH=CHCO2C2H5
Quantity 1 kg
Beilstein 02,742
Specific Gravity 1.05
Solubility Information Solubility in water: immiscible.
Formula Weight 172.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl fumarate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-819-7
RTECSNumber EM5950000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.