Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl maleate, ≥96.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013067 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3067 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3067 Supplier MilliporeSigma Supelco Supplier No. 075531ML
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 662°F
CAS 141-05-9
Vapor Pressure 1 mmHg (14°C)
Vapor Density 5.93 (vs. air)
Density 1.064 g/mL (at 25°C)
Flash Point 93°C
Linear Formula C2H5 OCOCH=CHCOOC2H5
MDL Number MFCD00009191
Melting Point -10°C
Boiling Point 225°C
Synonym Maleic acid diethyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 172.18
Percent Purity ≥96.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.