Learn More
Sigma Organic Chemistry Diethyl malonate 2 | 500G | 105-53-3 | MFCD00009195
Supplier: Sigma Organic Chemistry D97754500G

Diethyl malonate, 500G
About This Item:
Synonym(s): Malonic acid diethyl ester
Linear Formula: CH2(COOC2H5)2
Molecular Weight: 160.17
CAS Number: 105-53-3
MDL Number: MFCD00009195
UNSPSC Code: 12352100
Purity: 0.99
Boiling Point: 199 C (lit.)
Melting Point: -51--50 C (lit.)
Density: 1.055 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.413 (lit.)
Storage: room temp
EINECS Number: 203-305-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.