Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl malonate, 99%

Catalog No. p-7022609
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1546836 500 g
AAA154680E 2500 g
AAA154680C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1546836 Supplier Thermo Scientific Chemicals Supplier No. A1546836
Only null left
Add to Cart

CAS: 105-53-3 | C7H12O4 | 160.169 g/mol

Diethyl malonate is used in organic synthesis for the preparation of alpha-aryl malonates, mono-substituted and di-substituted acetic acid, barbiturates and artificial flavorings. It is also involved in the synthesis of pharmaceuticals like chloroquine, butazolidin and barbital. It acts as intermediate in the synthesis of vitamin B1, vitamin B6, non-steroidal anti-inflammatory agents agrochemicals and perfumes. In Knoevenagel condensation reaction, it reacts with benzaldehyde to get diethyl benzylidenemalonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl malonate is used in organic synthesis for the preparation of alpha-aryl malonates, mono-substituted and di-substituted acetic acid, barbiturates and artificial flavorings. It is also involved in the synthesis of pharmaceuticals like chloroquine, butazolidin and barbital. It acts as intermediate in the synthesis of vitamin B1, vitamin B6, non-steroidal anti-inflammatory agents agrochemicals and perfumes. In Knoevenagel condensation reaction, it reacts with benzaldehyde to get diethyl benzylidenemalonate.

Solubility
Miscible with ethyl alcohol, ether, chloroform and benzene. Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-53-3
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00009195
InChI Key IYXGSMUGOJNHAZ-UHFFFAOYSA-N
Synonym diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
PubChem CID 7761
IUPAC Name diethyl propanedioate
SMILES CCOC(=O)CC(=O)OCC

Specifications

Melting Point °C to 50°C
Density 1.055
Boiling Point 198°C to 200°C
Flash Point 90°C (194°F)
Assay Percent Range 99%
Refractive Index 1.414
Quantity 500 g
Beilstein 774687
Merck Index 14,3823
Solubility Information Miscible with ethyl alcohol,ether,chloroform and benzene. Slightly miscible with water.
Formula Weight 160.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl Malonate
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-305-9
RTECSNumber OO0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.