Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl oxalate, 99%

Catalog No. AAA144980E
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA144980E 2500 g
AAA1449836 500 g
AAA144980C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA144980E Supplier Thermo Scientific Chemicals Supplier No. A144980E
Add to Cart

CAS: 95-92-1 | C6H10O4 | 146.142 g/mol

Diethyl oxalate is used to prepare active pharmaceutical ingredients (API), plastics and dyestuff intermediates. It is also used as a solvent for cellulose esters, ethers, resins, perfumes and lacquers for electronics. It is involved in the transesterification reaction with phenol to get dipheny oxalate. It is also involved in the Claisen condensation ketosteroids to prepare glyoxalyl derivatives. Further, it is used to prepare sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine. In addition to this, it is utilized in the microemulsion synthesis of zinc oxide nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl oxalate is used to prepare active pharmaceutical ingredients (API), plastics and dyestuff intermediates. It is also used as a solvent for cellulose esters, ethers, resins, perfumes and lacquers for electronics. It is involved in the transesterification reaction with phenol to get dipheny oxalate. It is also involved in the Claisen condensation ketosteroids to prepare glyoxalyl derivatives. Further, it is used to prepare sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine. In addition to this, it is utilized in the microemulsion synthesis of zinc oxide nanoparticles.

Solubility
Miscible with alcohols, ether and other common organic solvents.

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-92-1
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00009119
InChI Key WYACBZDAHNBPPB-UHFFFAOYSA-N
Synonym ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove
PubChem CID 7268
IUPAC Name diethyl oxalate
SMILES CCOC(=O)C(=O)OCC

Specifications

Melting Point -41°C
Density 1.077
Boiling Point 184°C to 186°C
Flash Point 75°C (167°F)
Refractive Index 1.41
Quantity 2500 g
UN Number UN2525
Beilstein 606350
Sensitivity Moisture sensitive
Merck Index 14,3125
Solubility Information Miscible with alcohols,ether and other common organic solvents.
Formula Weight 146.14
Percent Purity 99%
Chemical Name or Material Diethyl oxalate
Show More Show Less
Hazard Category H227-H302-H319
Hazard Statement GHS H Statement
H302-H319-H227
Harmful if swallowed.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ETHYL OXALATE
EINECSNumber 202-464-1
RTECSNumber RO2800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.