Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl oxalpropionate

Catalog No. 137766100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-776-6100G 100 g
13-776-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-776-6100G Supplier Sigma Aldrich Supplier No. 137766100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant involved in:
  • Synthesis of anticancer and antiviral agents
  • Oxidation by organohypervalent iodine reagent
  • Synthesis of monomers for preparation of functional polyesters
  • Synthesis of calpain inhibitors
  • Preparation of human a2a receptor antagonists
  • Structural studies of dihydropteroate synthase inhibitors

Specifications

CAS 759-65-9
Density 1.073 g/mL (at 25°C (literature))
Boiling Point 138°C (23 mmHg)
Molecular Formula C9H14O5
Refractive Index n20/D 1.432 (literature)
Linear Formula C2H5OCOCH(CH3)COCOOC2H5
MDL Number MFCD00009163
Quantity 100 g
Synonym Diethyl 2-methyl-3-oxosuccinate; Methyloxo-butanedioic acid diethyl ester; NSC 33946
Molecular Weight (g/mol) 202.2
Percent Purity ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.