Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Phthalate, NF, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602191 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-191 4 L Amber Glass Bottle
18-602-192 20 L Amber Glass Bottle
18-602-193 200 L Amber Glass Bottle
18-602-190 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-191 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D10974LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

D1097, 84-66-2, C12H14O4

Spectrum™ Chemical Diethyl Phthalate, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
IUPAC Name 1,2-diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

CAS Max % 100%
Ignition Residue 0.02%
Packaging Amber Glass Bottle
Refractive Index 1.500-1.505
Quantity 4 L
Specific Gravity 1.118-1.122
Percent Purity 98 to 102%
Grade NF
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.