Learn More
Sigma Organic Chemistry Diethyl sebacate | 100ML | 110-40-7 | MFCD00009219 | 0.98
Supplier: Sigma Organic Chemistry 246077100ML

Diethyl sebacate, 100ML
About This Item:
Synonym(s): Decanedioic acid 1,10-diethyl ester; NSC 8911; Sebacic acid diethyl ester
Linear Formula: C2H5OCO(CH2)8COOC2H5
Molecular Weight: 258.35
CAS Number: 110-40-7
MDL Number: MFCD00009219
UNSPSC Code: 12162002
Purity: 0.98
Boiling Point: 312 C (lit.)
Melting Point: 1-2 C (lit.)
Density: 0.963 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.436 (lit.)
Storage: room temp
EINECS Number: 203-764-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.