Learn More
Sigma Organic Chemistry Diethyl sulfate | 2KG | 64-67-5 | MFCD00009099
Supplier: Sigma Organic Chemistry D1007062KG

Diethyl sulfate, 2KG
About This Item:
Synonym(s): Ethyl sulfate; Sulfuric acid diethyl ester
Linear Formula: (C2H5O)2SO2
Molecular Weight: 154.18
CAS Number: 64-67-5
MDL Number: MFCD00009099
UNSPSC Code: 12352108
Purity: 0.98
Boiling Point: 208 C (lit.)
Melting Point: -24 C (lit.)
Density: 1.177 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.399 (lit.)
Storage: room temp
EINECS Number: 200-589-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.