Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl sulfide, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019223 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9223 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9223 Supplier MilliporeSigma Supelco Supplier No. 417361ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 352-93-2
Vapor Pressure 105 mmHg (37.7°C)
Density 0.837 g/mL (at 25°C)
Linear Formula (C2H5)2 S
MDL Number MFCD00009270
Melting Point -100°C
Boiling Point 90°C to 92°C
Synonym 1,1 ′-Thiobisethane; Ethyl sulfide; NSC 75157
Type Analytical Standard
Quantity 1 mL
Formula Weight 90.19
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.