Diethylamine, ≥99%, Alfa Aesar™

Manufacturer: Alfa Aesar A117160F

Quantity 2500mL

 View more versions of this product

Catalog No. AAA117160F

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 109-89-7
    Physical Form Liquid
    Assay Percent Range ≥99%
    Molecular Formula (CH3CH2)2NH
    Formula Weight 73.14
    Chemical Name or Material Diethylamine

    Specifications

    Formula Weight 73.14
    Quantity 2500mL
    Physical Form Liquid
    Density 0.706
    Assay Percent Range ≥99%
    Molecular Formula (CH3CH2)2NH
    CAS 109-89-7
    Storage Note 1 Air Sensitive
    Chemical Name or Material Diethylamine

    Description

    Applications
    Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals. It is also used to cook lysergic acid diethylamide (LSD) and is strictly watched by Drug Enforcement Administration (DEA).

    Solubility
    Miscible with water, alcohol, ether, chloroform and carbon terachloride.

    Notes
    Air sensitive. Incompatible with strong oxidizing agents.

    Air Sensitive

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit