Learn More
AdipoGen Diethylaminocoumarin-3 COOH im
SDP
Supplier:  AdipoGen CDXD0071M100
Encompass_Preferred
Chemical. CAS 261943-47-9. Formula C17H17N3O3. MW 311.34. Synthetic. fluorescent reagent for use in circular dichroic exciton method for derivatisation of hydroxyl-groups under mild conditions. The absorption at 406 nm is futher in the red than for reagents like naphthoylimidazole. The reagent offers high sensitivity through a high molar extinction coefficient.
Catalog No. 50-256-1263
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.