Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylbenzene, mixture of isomers

Catalog No. AAA195480F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA195480F 2500 mL
AAA19548AE 100 mL
AAA19548AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA195480F Supplier Thermo Scientific Chemicals Supplier No. A195480F
Add to Cart

CAS: 25340-17-4 | C10H14

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25340-17-4
Molecular Formula C10H14
MDL Number MFCD00792900
Synonym Dowex J

Specifications

Density 0.87
Boiling Point 182°C to 184°C
Flash Point 55°C (131°F)
Refractive Index 1.495
Quantity 2500 mL
UN Number UN2049
Beilstein 1903396
Formula Weight 134.22
Chemical Name or Material Diethylbenzene, mixture of isomers
Hazard Category H226-H304-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber 246-874-9
RTECSNumber CZ5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.